Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

1,2-Dioctanoyl-sn-glycerol (8:0), Calbiochem®

Supplier: MilliporeSigma

317505-10MG
80051-434EA 94.89 USD InStock
80051-434
1,2-Dioctanoyl-sn-glycerol (8:0), Calbiochem®
1,2-Dioctanoyl-sn-glycerol (8:0)
1,2-Dioctanoyl-sn-glycerol is a cell-permeable activator of protein kinase C (PKC)

Shown to mimic the effects of tumor-promoting phorbol diesters on mitogenesis and epidermal growth factor binding and action in intact cells.

Exhibits lower affinity for PKCα than for other isozymes.


Formula: C₁₉H₃₆O₅
CAS Number: 60514-48-9
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty