Would you like to log in again?
SP6 RNA Polymerase, Promega
Supplier: Promega
P1085 P1081
PAP1085EA 330.54 USD InStock
PAP1085 PAP1081
SP6 RNA Polymerase, Promega
Nucleic Acid Reagents RNA Synthesis and Transcription
DNA-dependent RNA polymerase for in vitro transcription.
  • DNA-Dependent RNA Polymerase Specific for SP6 Promoter Sequences
  • Exhibits extremely high affinity and specificity for SP6 promoter sequences
  • >90% pure as determined by SDS polyacrylamide gel electrophoresis
  • Free of detectable levels of contaminating RNase and DNase activity (
SP6 RNA Polymerase is a DNA-dependent RNA polymerase that exhibits extremely high specificity for its cognate promoter sequences. Only SP6 DNA or DNA cloned downstream from an SP6 promoter can serve as a template for SP6 RNA Polymerase-directed RNA synthesis.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty