Would you like to log in again?
BBL™ Blood Agar Slants, BD Diagnostics
Supplier: BD
BBL™
220830 220830
90001-132PK 161.84 USD InStock
90001-132 CA90001-132
BBL™ Blood Agar Slants, BD Diagnostics
Media for Microbiology Prepared Media for Microbiology
Blood Agar is an enriched medium for the isolation and growth of fastidious microorganisms and for the determination of hemolytic reactions.
  • Casein and soy peptones provide essential nitrogen
  • Sodium chloride maintains osmotic equilibrium
  • Growth of Haemophilus hemolyticus is inhibited
The nutritional composition of Trypticase Soy Agar has made it a popular medium, both unsupplemented and as a base for media containing blood. Trypticase Soy Agar with 10% Sheep Blood is used for the recovery and cultivation of fastidious microbial species and for the determination of hemolytic reactions which are important differentiating characteristics for bacteria, especially Streptococcus species.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty