Would you like to log in again?
BBL™ Chocolate Agar Slants, BD Diagnostics
Supplier: BD
BBL™
295872
90004-604PK 122.93 USD InStock
90004-604
BBL™ Chocolate Agar Slants, BD Diagnostics
Media for Microbiology Prepared Media for Microbiology
Chocolate Agar is recommended for the cultivation and isolation of Neisseria and Haemophilus species.
  • Casein and meat peptones provide essential nitrogen
  • Phosphate buffers maintain pH level
  • Corn starch neutralizes toxic fatty acids
Chocolate agar contains a GC agar base, bovine hemoglobin, and a chemically defined treatment. This product is for in vitro diagnostic use only.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty