Would you like to log in again?
BBL™ Chopped Meat Carbohydrate Broth, BD Diagnostics
Supplier: BD
BBL™
297307
90004-832PK 168.78 USD InStock
90004-832
BBL™ Chopped Meat Carbohydrate Broth, BD Diagnostics
Media for Microbiology Prepared Media for Microbiology
Chopped Meat Carbohydrate Broth, PR II (pre-reduced medium in Hungate tubes) is a general-purpose medium for the enrichment and cultivation of anaerobic microorganisms, particularly obligate anaerobes.
  • Anaerobic nitrogen and hydrogen atmosphere
  • Packaged under oxygen-free conditions
  • Meat pellets and enzymatic digest of casein provide nitrogen
  • Yeast extract provides B-complex vitamins
  • Glucose is a source of energy
  • Phosphate maintains the pH of the medium
The reducing action of the meat particles and L-cysteine binds molecular oxygen. The reducing agents are required to maintain a low Eh. Resazurin is an oxidation-reduction indicator used to detect changes in the Eh of the medium. The medium remains colorless if the Eh remains low; increased oxidation causes the medium to become pink.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty