Would you like to log in again?
Difco™ Modified Oxford Antimicrobic Supplement, BD Diagnostics
Supplier: BD Diagnostics
BBL™
211763 211763
90000-336PK 565.63 USD InStock
90000-336 CA90000-336
Difco™ Modified Oxford Antimicrobic Supplement, BD Diagnostics
Media for Microbiology Prepared Media for Microbiology
Oxford Medium Base is a selective, solid medium used with Oxford Antimicrobic Supplement or Modified Oxford Antimicrobic Supplement for isolating and differentiating Listeria monocytogenes in foods.
  • Recommended for isolating and identifying Listeria monocytogenes
  • Used with processed meat and poultry products
  • Improves selectivity of Oxford Medium Base
Incorporating this agent into Oxford Medium Base will completely inhibit gram-negative organisms and most gram-positive organisms after 24 hours of incubation.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty