Would you like to log in again?
Difco™ MIO Medium, BD Diagnostics
Supplier: BD Diagnostics
Difco™
273520 273520
90003-908EA 304.89 USD InStock
90003-908 CA90003-908
Difco™ MIO Medium, BD Diagnostics
Media for Microbiology Prepared Media for Microbiology
Motility Indole Ornithine (MIO) Medium is a semisolid medium useful in the identification of members of the Enterobacteriaceae.
  • Casein and peptic digest provide nitrogen and amino acids
  • Yeast extract is a source of vitamin B-complex
  • Dextrose is the fermentable carbohydrate
Motility Indole Ornithine Medium was formulated by Ederer and Clark and Oberhofer and Hajkowski for detection of motility, indole and ornithine decarboxylase production in one tube as an aid in the identification of members of the Enterobacteriaceae family.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty