Would you like to log in again?
Accessories for ETS-D5 Electronic Contact Thermometer, IKA
Supplier: IKA
0002735600
33993-812EA 239.33 USD InStock
33993-812
Accessories for ETS-D5 Electronic Contact Thermometer, IKA
Thermometers Digital Thermometers
Extension cable to separate the casing from the sensor. The casing with the electronics may thus be kept away from dangerous vapor released by the medium (for use with ETS-D5 and ETS-D6).
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty