Would you like to log in again?
Microchem® by AlphaTec™ 68-6000 Gas-Tight Chemical Protection Coveralls, Ansell
Supplier: Ansell
OR60-T-92-803-05-GA1 OR60-T-92-803-07-GA1 OR60-T-92-803-02-GA1 OR60-T-92-802-05-GA1 OR60-T-92-801-06-G02 OR60-T-92-801-04-G02 OR60-T-92-803-04-GA1 OR60-T-92-802-04-GA1 OR60-T-92-801-03-G02 OR60-T-92-801-02-G02 OR60-T-92-803-06-GA1 OR60-T-92-803-03-GA1 OR60-T-92-802-02-GA1 OR60-T-92-802-03-GA1 OR60-T-92-802-06-GA1 OR60-T-92-802-07-GA1 OR60-T-92-801-07-G02 OR60-T-92-801-05-G02
10836-334CS 5696.43 USD InStock
10836-334 10836-312 10836-322 10836-498 10836-310 10836-332 10836-320 10836-496 10836-316 10836-326 10836-314 10836-324 10836-308 10836-318 10836-306 10836-330 10836-494 10836-492
Microchem® by AlphaTec™ 68-6000 Gas-Tight Chemical Protection Coveralls, Ansell
Disposable Garments
MICROCHEM® by AlphaTec™ 6000 is a range of lightweight, flexible and strong Level A Gas-Tight suits designed to be worn with self contained breathing apparatus to provide protection for emergency responders or chemical workers against dangerous and toxic chemicals in either liquid or gaseous form.
  • Tested against numerous chemicals including the ASTM F1001 test battery
  • Co-extruded multi-layer performance barrier laminate with scrim reinforcement
  • Design features include a DYNAT/YKK Gas-Tight zipper that provides protection and performance in the most hostile of chemical environments
  • Includes semi-rigid 3-layer PET visor providing excellent optical clarity
Disposable latex suits with ultrasonically welded and taped seams. Typically used in chemicals, pharmaceuticals, fire service, sewage treatment, nuclear, petrochemicals, first response, industrial and tank cleaning, and health service shipping.
Certifications: Tested according to EN 1073-2, EN1149-5 / AATCC 76, EN 14126 / ASTM F1671, EN943-1 Type 1a, EN943-2 Type 1a-ET.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty