Would you like to log in again?
Whatman™ Activated Carbon Air Filter, Grade 72, Whatman products (Cytiva)
Supplier: Whatman products (Cytiva)
Whatman™ HyClone™
1872-055 1872-050 1872-055 1872-047 1872-047 1872-050
76324-946PK 120.4 USD InStock
76324-946 89187-086 CA76333-742-1 CA89187-086 76423-734 CA76423-734
Whatman™ Activated Carbon Air Filter, Grade 72, Whatman products (Cytiva)
Filters Filter Paper Qualitative Filter Paper
Grade 72 activated carbon air filter paper is designed for air pollution monitoring of radioactive iodine in medical and nuclear installations.
  • High-quality composite cellulose/glass filter paper
  • Loaded with activated carbon for radioactive iodine absorption
  • Suitable for use in radioactive environments
Air pollution monitoring of radioactive iodine in medical or nuclear installations is a critical safety requirement. Grade 72 cellulose filter papers are designed specifically for these environments, delivering sensitive and accurate results by absorbing any radioactive iodine in the air for analysis.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty