Would you like to log in again?
Econowipe Cleanroom Wipes, Liberty Industries
Supplier: Liberty Industries
50-5120
21924-470CS 647.22 USD InStock
21924-470
Econowipe Cleanroom Wipes, Liberty Industries
Wipes Cleanroom Wipes Dry Cleanroom Wipes
These cleanroom wipes are perfect for light to medium duty wiping applications.
  • Will not break down when wet
  • Will absorb many times its own weight in liquid
Packaged in sturdy cleanroom dispenser plastic tubs that are ideal for clean storage and easy use. Monitored and tested to insure low levels of particulate matter, sodium and other heavy metals.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty