Would you like to log in again?
Disposable Bags, Polyethylene, Brady
Supplier: Brady
145587
10796-478EA 575.64 USD InStock
10796-478
Disposable Bags, Polyethylene, Brady
Bags Waste Disposal Bags
These disposal bags are made of durable 3 mil polyethylene.
Ideal for securely store, ship and dispose of infectious materials as OSHA requires.
Packaging: 100 bags per pack.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty