Would you like to log in again?
Whatman™ Solvent IFD In-Line Filter/Degassers
Supplier: Whatman products (Cytiva)
Whatman™
6725-5002A 6725-5002 6725-5002 6725-5002A
28137-971PK 254.6 USD InStock
CA11013-008 28137-971 CA28137-971 10035-492
Whatman™ Solvent IFD In-Line Filter/Degassers
Filters In-Line Filters
Cytiva's Solvent IFD is a high-flow HPLC in-line filter/degasser, designed to simultaneously filter and degas organically based HPLC mobile phases.
  • Faster than traditional mobile phase preparation
  • Chemically resistant polypropylene membrane and housing, built to filter solvent mobile phases without expensive stand-alone degassing equipment
  • Air vent on inlet with luer lock cap
  • Integrity-testable with the bubble point method
  • End fittings included
Whatman IFD options from Cytiva's business are available for a variety of applications.
A sturdy and chemically resistant medium is needed to filter and degas an organic solvent. Polypropylene is largely inert, compatible with many chemicals, and retains stability across a range of temperatures. The 0.2 µm high-flow polypropylene membrane and housing makes Solvent IFD suitable to filter and degas these solvents.
The Solvent IFD’s all-polypropylene housing is sealed by a security ring. The filter includes fittings to accommodate ¹/₁₆ to ¹/₈" tubing and an air vent on the inlet with luer lock cap to enable priming.
The in-line filter works on the principle of 'bubble point' – the point of pressure at which gases will pass through a wet membrane. If pressure is maintained below the bubble point, the gas will not pass through the membrane and is trapped by the particular filter device.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty