Would you like to log in again?
Tergitol™ NP-9, Surfactant, Spectrum
Supplier: Spectrum
Tergitol™
T1285-500ML T1285-20LT T1285-4LT T1285-200LTB T1285-100ML
700011-160EA 2474 USD InStock
700011-160 700011-158 700011-166 700011-162 700011-164
Tergitol™ NP-9, Surfactant, Spectrum
Cleaning Agents Detergents and Surface Cleansers
Tergitol™ NP-9, Surfactant is a Nonylphenol Ethoxylate based surfactant. It is nonionic and provides excellent detergency, outstanding wetting, versatile solubility characteristics and low odor. This product is suggested for use in cleaners & detergents, prewash spotters, paints & coatings, dust control and metalworking fluids.
  • Delivers a combination of economy and performance
  • Excellent detergency and wetting
  • Good solubilization and emulsification
Density at 20°C (68°F): 1.055 g/ml
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty