Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Calbiochem® Human Albumin

Supplier: EMD Millipore
Calbiochem®
12667-50ML
80055-732EA 632.84 USD InStock
80055-732
Calbiochem® Human Albumin
Proteins and Peptides
Purified plasma protein that is useful as a stabilizing agent and as a carrier protein.

  • Molar Mass: 66000
  • Purity: ≥ 95% by agarose electrophoresis
  • Toxicity: Standard Handling
  • Storage: +2 to +8 °C

Prepared from serum that has been shown by certified tests to be negative for HBsAg and for antibodies to HIV and HCV.

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty