Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Taurodeoxycholic acid sodium salt ≥95% (by TLC), Calbiochem®, Millipore®

Supplier: MilliporeSigma
Synonyms: Cyclopenta[a]phenanthren-17-yl)pentanamido)ethanesulfonateCyclopenta[a]phenanthren-17-yl)pentanamido)ethanesulphonateSodium taurodeoxylateTaurodeoxycholate sodium salt

M-Clarity™ quality level = MQ100

580221-50GM 580221-5GM 580221-5GM
80058-712EA 770.9 USD InStock
80058-712 CA80058-714 80058-714
Taurodeoxycholic acid sodium salt ≥95% (by TLC), Calbiochem®, Millipore®
Taurodeoxycholic acid sodium salt
Taurodeoxycholic acid is a detergent useful for the solubilization of lipids and membrane-bound proteins.

  • White solid
  • CMC: 1 to 4 mM
  • Aggregation number: 6


Formula: C₂₆H₄₄NO₆S·Na
MW: 521.7 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00003671
CAS Number: 1180-95-6
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty