Would you like to log in again?
X-Gluc solution, Novagen® colorimetric stain, Molecular biology grade, Sigma-Aldrich®
Supplier: MilliporeSigma
70036-3
EM70036-3EA 216.43 USD InStock
EM70036-3
X-Gluc solution, Novagen® colorimetric stain, Molecular biology grade, Sigma-Aldrich®
X-Gluc solution
The β-glucuronidase substrate X-Gluc (5-bromo-4-chloro-3-indolyl-β-D-glucuronide) is a colorimetric stain for GUS activity and is used to identify recombinants containing pBACgus vectors, and LIC GUS Control Insert.
  • Colorimetric stain
  • Used in staining plaques for GUS activity
X-Gluc Solution is used in staining plaques for GUS activity. Each vial contains enough substrate to stain 15 100 mm plates.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty