Would you like to log in again?
Readycult®, Coliforms 100, MilliporeSigma
Supplier: MilliporeSigma
Readycult®
1.01298.0001
EM1.01298.0001EA 310.97 USD InStock
EM1.01298.0001
Readycult®, Coliforms 100, MilliporeSigma
Media for Microbiology Prepared Media for Microbiology
Readycult® Coliforms 100 is a selective enrichment broth.
  • Simultaneous detection
  • Total coliforms and E. coli within the bacteriological water examination
  • Selective enrichment broth
Readycult® Coliforms 100 is a selective enrichment broth for the simultaneous detection of total coliforms and E. coli within the bacteriological water examination.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty