Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Scheduled maintenance of the site is planned from June 8th at 7:00 PM to June 9th at 5:00 PM ET. During this time, contract pricing and registration functionality will be unavailable. We appreciate your patience.

Water ≥99%, CHROMASOLV™ for pesticide residue analysis, Riedel-de Haën™

Supplier: Honeywell Research Chemicals
Synonyms: Dihydrogen oxideIce

34478-6X1L
BJ34478-1LCS 732.43 USD InStock
BJ34478-1L
Water ≥99%, CHROMASOLV™ for pesticide residue analysis, Riedel-de Haën™
Water
High purity Water; suitable for Pesticide Residue Analysis (PRA) applications. Contains no preservative.

  • For gas chromatography (GC)
  • Developed especially for the application in residue analysis of pesticides and other low-volatile substances using GC
  • Non-volatile matter and water content are specified for each solvent


Formula: H₂O
MW: 18.02 g/mol
Boiling Pt: 100 °C (1.013 hPa)
Melting Pt: 0 °C
Density: 1 g/cm³ (20 °C) (lit.)
Storage Temperature: Ambient
MDL Number: MFCD00011332
CAS Number: 7732-18-5
EINECS: 231-791-2
Order Now
Specification Test Results
Non-volatile matter Max. 0.0005 %
Suitability for residue analysis Max. 5 ng/l
Determination of P/N ≤5 ng/L

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty