Would you like to log in again?
Malt Extract Powder, Refined, HiMedia Laboratories
Supplier: HiMedia Laboratories
RM004B-500G
61001-518EA 114.63 USD InStock
61001-518
Malt Extract Powder, Refined, HiMedia Laboratories
Media for Microbiology Prepared Media for Microbiology
Employed for cultivation of fungi such as malt agar, malt extract agar and yeast malt agar.
  • Cultivation of fungi
  • Malt extract powder
  • Sutiable for use in media
This malt extract powder is rich in carbohydrates and is ideally suitable for use in media. Employed for cultivation of fungi such as malt agar, malt extract agar and yeast malt agar.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty