Would you like to log in again?
Due to scheduled system maintenance, vwr.com will be temporarily unavailable from 6:00 am EST to 2:00 PM EST on Saturday, May 25. 
Berm Repair Kits, Justrite®
Supplier: Justrite
28330 28328
10861-262EA 411.34 USD InStock
10861-262 10861-264
Berm Repair Kits, Justrite®
Spill Control Materials
Use to repair berms having small holes, abrasion points, and small tears to maintain fluid-tight reliability and compliance.
  • Used to repair berms in the field, to help prolong life.
Contains heat gun and nozzle, pressure application roller, sand paper, scissors and utility knife, PVC cement, and 10-square feet of coated fabric repair material.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty