Would you like to log in again?
White Blood Cell System, HemoCue
Supplier: HemoCue
HemoCue®
113003 W1PROMO 123022 WBC00S
10842-820EA 2359.99 USD InStock
10842-820 75927-794 10842-828 75927-778
White Blood Cell System, HemoCue
Clinical Diagnostic Supplies Clinical Analysers
Based on HemoCue’s proven, reliable microcuvette technology, the HemoCue WBC System brings you the unique benefit of determining total white blood cell counts right at the point of care
  • Lab Accurate
  • Powered by AC power or 4 AA Batteries
  • Results within 3 minutes
  • Small Sample Size - 10 ul
  • Measurement Range 300 - 30,000/uL
  • Capillary or Venous Blood
Fitting seamlessly into a variety of clinical applications and even remote field clinics, the benefits are clear. Immediate WBC counts can mean the difference between waiting and taking action at the point of care—helping you move from assessment to treatment within minutes rather than hours or days.
In just three minutes, you have results with the same precision and accuracy as those from a central laboratory.
Individual cuvette (sold separately) stability: Unopened 1 Year; Opened: 3 months
Certifications: Moderately Complex.
Ordering information: Analyzer and cuvettes sold separately.
Order Now
ORDER
ACCESSORIES
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty