Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

N-Cbz-N'-boc-L-ornithine

Supplier: INDOFINE CHEMICAL COMPANY MS
Synonyms: Z-Orn(Boc)-OH(S)-2-(((Benzyloxy)carbonyl)amino)-5-((tert-butoxycarbonyl)amino)pentanoic acidNα-Benzyloxycarbonyl-Nδ-Boc-L-ornithine

048275GM
100270-564EA 329.54 USD InStock
100270-564
N-Cbz-N'-boc-L-ornithine
N-Cbz-N'-boc-L-ornithine


Formula: C₁₈H₂₆N₂O₆
MW: 366.414 g/mol
MDL Number: MFCD00038268
CAS Number: 7733-29-1
ADR: ,
https://us-legacy.vwr.com/stibo/web/std.lang.all/59/82/21285982.jpg
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty