Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2-Bromethylamine hydrobromide ≥98%

Supplier: INDOFINE CHEMICAL COMPANY MS
Synonyms: 2-Bromoethylamine hydrobromide2-Bromoethylammonium bromide

20101100GM
100267-750EA 52.31 USD InStock
100267-750
2-Bromethylamine hydrobromide ≥98%
2-Bromethylamine hydrobromide


Formula: BrCH₂CH₂NH₃Br
MW: 204.89 g/mol
Melting Pt: 172…174 °C
MDL Number: MFCD00012886
CAS Number: 2576-47-8
ADR: ,
https://us-legacy.vwr.com/stibo/web/std.lang.all/57/69/21285769.jpg
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty