Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Capsaicin ≥95% (by HPLC)

Supplier: INDOFINE CHEMICAL COMPANY MS
Synonyms: 8-Methyl-N-vanillyl-6-none-amid

02898610MG
100269-068EA 120.31 USD InStock
100269-068
Capsaicin ≥95% (by HPLC)
Capsaicin


Formula: C₁₈H₂₇NO₃
MW: 305.42 g/mol
MDL Number: MFCD00017259
CAS Number: 404-86-4
UN: 2811
ADR: 6.1,III
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty