Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Iota carrageenan, powder

Supplier: INDOFINE CHEMICAL COMPANY MS

02437550GM
100266-284EA 298.15 USD InStock
100266-284
Iota carrageenan, powder
Iota carrageenan
  • Light yellow powder
  • Store in a cool, dry place


MDL Number: MFCD00151512
CAS Number: 9062-07-1
ADR: ,
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty