Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

(S)-2-Amino-2-cyclohexylacetic acid, Powder

Supplier: INDOFINE CHEMICAL COMPANY MS
Synonyms: H-Chg-OH(S)-alpha-Aminocyclohexylacetic AcidH-Cyclohexyl-Gly-OH

0410651GM
100270-918EA 85 USD InStock
100270-918
(S)-2-Amino-2-cyclohexylacetic acid, Powder
(S)-2-Amino-2-cyclohexylacetic acid
  • White powder
  • Store at 0-8 °C


Formula: C₈H₁₅NO₂
MW: 157.2 g/mol
MDL Number: MFCD01311679
CAS Number: 14328-51-9
ADR: ,
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty