Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

α,α,α-Trifluoro-3,4-diaminotoluene ≥98%, powder

Supplier: INDOFINE CHEMICAL COMPANY MS
Synonyms: 4-(Trifluoromethyl)benzene-1,2-diamine4-(Trifluoromethyl)-1,2-phenylenediamine4-(Trifluoromethyl)-o-phenylenediamine3,4-Diaminobenzotrifluoride

097195GM
100283-814EA 109.85 USD InStock
100283-814
α,α,α-Trifluoro-3,4-diaminotoluene ≥98%, powder
α,α,α-Trifluoro-3,4-diaminotoluene
  • Beige powder
  • Store in a cool, dry place


Formula: C₇H₇F₃N₂
MW: 176.14 g/mol
Melting Pt: 58…60 °C
MDL Number: MFCD00042456
CAS Number: 368-71-8
UN: 2811
ADR: 6.1,III
https://us-legacy.vwr.com/stibo/web/std.lang.all/68/35/21286835.jpg https://us-legacy.vwr.com/stibo/web/std.lang.all/68/36/21286836.jpg
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty