Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Sinapic acid ≥99%, Powder

Supplier: INDOFINE CHEMICAL COMPANY MS
Synonyms: 4-Hydroxy-3,5-dimethoxycinnamic acid3',5'-Dimethoxy-4'-hydroxycinnamic acid3,5-Dimethoxy-4-hydroxycinnamic acid

0113545GM
100265-812EA 52.31 USD InStock
100265-812
Sinapic acid ≥99%, Powder
Sinapic acid
  • Pale yellow powder
  • Store in cool dry place


Formula: C₁₁H₁₂O₅
MW: 224.21 g/mol
Melting Pt: 200…203 °C
MDL Number: MFCD00004401
CAS Number: 530-59-6
ADR: ,
https://us-legacy.vwr.com/stibo/web/std.lang.all/56/70/21285670.jpg
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty