Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Escin 90-95%, Powder

Supplier: INDOFINE CHEMICAL COMPANY MS
Synonyms: Aescin

0201151GM
100263-374EA 176.54 USD InStock
100263-374
Escin 90-95%, Powder
Escin
  • White powder
  • Store in a Dry and Dark Place


Formula: C₅₅H₈₆O₂₄
MW: 1131.26 g/mol
Melting Pt: ∼221 °C
MDL Number: MFCD00076054
CAS Number: 6805-41-0
ADR: ,
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty