Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

3-Fluoro-p-anisaldehyde ≥98%

Supplier: INDOFINE CHEMICAL COMPANY MS
Synonyms: 3-Fluoro-4-anisaldehyde3-Fluoro-4-methoxybenzaldehyde

269575GM
100294-240EA 104.62 USD InStock
100294-240
3-Fluoro-p-anisaldehyde ≥98%
3-Fluoro-p-anisaldehyde


Formula: C₈H₇FO₂
MW: 154.14 g/mol
Melting Pt: 34…35 °C
MDL Number: MFCD00003349
CAS Number: 351-54-2
ADR: ,
https://us-legacy.vwr.com/stibo/web/std.lang.all/72/98/21287298.jpg
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty