Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

(R)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-3-(4-(tert-butoxy)phenyl)propanoic acid ≥99% (by HPLC), Powder

Supplier: INDOFINE CHEMICAL COMPANY MS
Synonyms: O-tert-Butyl-Nalpha-Fmoc-D-tyrosineFmoc-D-Tyr(tBu)-OHN-Fmoc-O-tert-butyl-D-tyrosineNalpha-Fmoc-O-tert-butyl-D-tyrosine

0412241GM
100278-908EA 122.92 USD InStock
100278-908
(R)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-3-(4-(tert-butoxy)phenyl)propanoic acid ≥99% (by HPLC), Powder
(R)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-3-(4-(tert-butoxy)phenyl)propanoic acid
  • White powder
  • Store at 0-8 °C


Formula: C₂₈H₂₉NO₅
MW: 459.5 g/mol
Melting Pt: 143…162 °C
MDL Number: MFCD00065684
CAS Number: 118488-18-9
ADR: ,
https://us-legacy.vwr.com/stibo/web/std.lang.all/74/79/21287479.jpg
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty