Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

D-(+)-Galactose ≥98% (by HPLC), crystalline powder

Supplier: INDOFINE CHEMICAL COMPANY MS
Synonyms: (2R,3S,4S,5R)-2,3,4,5,6-Pentahydroxyhexanal(2R,3S,4S,5R)-Pentahydroxyhexanal

024117100GM
100263-048EA 128.15 USD InStock
100263-048
D-(+)-Galactose ≥98% (by HPLC), crystalline powder
D(+)-Galactose
  • White crystalline powder
  • Store in a cool, dry place


Formula: C₆H₁₂O₆
MW: 180.16 g/mol
Melting Pt: 168…170 °C
MDL Number: MFCD00151230
CAS Number: 59-23-4
ADR: ,
https://us-legacy.vwr.com/stibo/web/std.lang.all/55/33/21285533.jpg
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty