Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

D-(-)-Glutamic acid ≥98.5%, crystalline powder

Supplier: INDOFINE CHEMICAL COMPANY MS
Synonyms: (R)-(-)-Glutamic acid(R)-(-)-1-Aminopropane-1,3-dicarboxylic acidD(-)-α-Glutamic acidH-D-Glu-OHD(-)-1-Aminopropane-1,3-dicarboxylic acid(R)-(-)-α-Glutamic acid(R)-2-Aminopentanedioic acid(R)-(-)-2-Aminopentanedioic acidD(-)-2-Aminopentanedioic acid

0255895GM
100263-752EA 35.31 USD InStock
100263-752
D-(-)-Glutamic acid ≥98.5%, crystalline powder
D(-)-Glutamic acid
  • White crystalline powder
  • Store in a cool, dry place


Formula: C₅H₉NO₄
MW: 147.13 g/mol
Melting Pt: 205 °C (dec.)
MDL Number: MFCD00063112
CAS Number: 6893-26-1
ADR: ,
https://us-legacy.vwr.com/stibo/web/std.lang.all/55/81/21285581.jpg
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty