Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Harmaline ≥98%, crystals

Supplier: INDOFINE CHEMICAL COMPANY MS
Synonyms: 3,4-Dihydro-7-methoxy-1-methyl-beta-carboline3,4-Dihydro-7-methoxy-1-methyl-9H-pyrido[3,4-b]indoleDihydroharmine

H0011GM
100265-698EA 28.77 USD InStock
100265-698
Harmaline ≥98%, crystals
Harmaline
  • Tan Crystals
  • Store at +4 °C


Formula: C₁₃H₁₄N₂O
MW: 214.27 g/mol
Melting Pt: 239…241 °C
MDL Number: MFCD00004955
CAS Number: 304-21-2
ADR: ,
https://us-legacy.vwr.com/stibo/web/std.lang.all/56/58/21285658.jpg
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty