Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

(R)-(-)-Mandelic acid ≥99% (by titrimetric analysis), crystalline powder

Supplier: INDOFINE CHEMICAL COMPANY MS
Synonyms: (R)-(-)-2-Hydroxy-2-phenylacetic acid(R)-2-Hydroxy-2-phenylacetic acidD(-)-2-Hydroxy-2-phenylacetic acidD(-)-Mandelic acid

041815100GM
100294-748EA 124.23 USD InStock
100294-748
(R)-(-)-Mandelic acid ≥99% (by titrimetric analysis), crystalline powder
(R)-(-)-Mandelic acid
  • White crystalline powder
  • Store at 0-8 °C


Formula: C₈H₈O₃
MW: 152.15 g/mol
Melting Pt: 129…135 °C
MDL Number: MFCD00064251
CAS Number: 611-71-2
ADR: ,
https://us-legacy.vwr.com/stibo/web/std.lang.all/73/54/21287354.jpg
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty