Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

5-Methoxypiperonal ≥97%

Supplier: INDOFINE CHEMICAL COMPANY MS
Synonyms: 7-Methoxybenzo[d][1,3]dioxole-5-carbaldehyde

0250481GM
100269-958EA 167.38 USD InStock
100269-958
5-Methoxypiperonal ≥97%
5-Methoxypiperonal


Formula: C₉H₈O₄
MW: 180.16 g/mol
Melting Pt: 129…132 °C
MDL Number: MFCD00016900
ADR: ,
https://us-legacy.vwr.com/stibo/web/std.lang.all/59/38/21285938.jpg
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty