Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Methyl-4-nitrobenzoate, crystalline powder

Supplier: INDOFINE CHEMICAL COMPANY MS
Synonyms: 4-Nitrobenzoic acid emthyl esterMethyl 4-nitrobenzoate

01116910GM
100262-922EA 49.69 USD InStock
100262-922
Methyl-4-nitrobenzoate, crystalline powder
Methyl 4-nitrobenzoate
  • Very pale greenish-yellow fine crystalline powder
  • Store in a cool, dry place


Formula: C₈H₇NO₄
MW: 181.15 g/mol
Melting Pt: 95…96 °C
MDL Number: MFCD00007350
CAS Number: 619-50-1
ADR: ,
https://us-legacy.vwr.com/stibo/web/std.lang.all/55/17/21285517.jpg
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty