Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Myricetin ≥99% (by HPLC), powder

Supplier: INDOFINE CHEMICAL COMPANY MS
Synonyms: 3,3',4',5,5',7-Hexahydroxyflavone3,5,7-Trihydroxy-2-(3,4,5-trihydroxyphenyl)-4H-chromen-4-one

021127S10MG
100266-078EA 228.85 USD InStock
100266-078
Myricetin ≥99% (by HPLC), powder
Myricetin
  • Powder
  • To be kept in a desiccator


Formula: C₁₅H₁₀O₈
MW: 318.24 g/mol
Melting Pt: 351 °C
MDL Number: MFCD00006827
CAS Number: 529-44-2
ADR: ,
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty