Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Nitro blue tetrazolium chloride (NBT) ≥99%, powder

Supplier: INDOFINE CHEMICAL COMPANY MS
Synonyms: 5,5'-Diphenyl-3,3'-bis(4-nitrophenyl)-2,2'-(3,3'-dimethoxybiphenyl-4,4'-ylene)ditetrazolium dichloride

MB1019100MG
100270-436EA 32.69 USD InStock
100270-436
Nitro blue tetrazolium chloride (NBT) ≥99%, powder
Nitro blue tetrazolium chloride (NBT)
  • Yellow powder
  • Store at +4 °C


Formula: C₄₀H₃₀Cl₂N₁₀O₆
MW: 817.65 g/mol
Melting Pt: 189 °C
MDL Number: MFCD00012159
CAS Number: 298-83-9
ADR: ,
https://us-legacy.vwr.com/stibo/web/std.lang.all/59/61/21285961.jpg https://us-legacy.vwr.com/stibo/web/std.lang.all/59/60/21285960.jpg
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty