Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

D-(-)-Serine ≥98% (by titrimetric analysis), crystalline powder

Supplier: INDOFINE CHEMICAL COMPANY MS
Synonyms: (R)-(-)-SerineH-D-Ser-OH

0256055GM
100266-418EA 35.31 USD InStock
100266-418
D-(-)-Serine ≥98% (by titrimetric analysis), crystalline powder
D(-)-Serine
  • White crystalline powder
  • Store in a cool, dry place


Formula: C₃H₇NO₃
MW: 105.09 g/mol
MDL Number: MFCD00004269
CAS Number: 312-84-5
ADR: ,
https://us-legacy.vwr.com/stibo/web/std.lang.all/57/13/21285713.jpg
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty