Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

n-Triacontane ≥99%

Supplier: INDOFINE CHEMICAL COMPANY MS

053000100MG
100277-532EA 258.92 USD InStock
100277-532
n-Triacontane ≥99%
n-Triacontane


Formula: H₃C(CH₂)₂₈CH₃
MW: 422.82 g/mol
MDL Number: MFCD00009410
CAS Number: 638-68-6
ADR: ,
https://us-legacy.vwr.com/stibo/web/std.lang.all/64/58/21286458.jpg
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty