Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

(R)-(-)-2-Amino-3-methyl-1-butanol, crystalline powder

Supplier: INDOFINE CHEMICAL COMPANY MS
Synonyms: (R)-(-)-2-Amino-3-methylbutanolD(-)-2-Amino-3-methyl-1-butanolD(-)-ValinolD-Valinol

0426871GM
100281-020EA 57.54 USD InStock
100281-020
(R)-(-)-2-Amino-3-methyl-1-butanol, crystalline powder
(R)-(-)-2-Amino-3-methyl-1-butanol
  • White crystalline powder
  • Store at 0-8 °C


Formula: C₅H₁₃NO
MW: 103.16 g/mol
MDL Number: MFCD00064297
CAS Number: 4276-09-9
ADR: ,
https://us-legacy.vwr.com/stibo/web/std.lang.all/74/99/21287499.jpg
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty