Would you like to log in again?
BBL™ Lauryl Tryptose Broth with MUG, BD Life Sciences
Supplier: BD Life Sciences
211744 211744
90000-318EA 743.78 USD InStock
90000-318 CA90000-318
BBL™ Lauryl Tryptose Broth with MUG, BD Life Sciences
Media for Microbiology Prepared Media for Microbiology
Broth with MUG Fluorogenic detection of E.coli in water, food and milk.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty