Would you like to log in again?
Maxwell® 16 Tissue LEV Total RNA Purification Kit, Promega
Supplier: Promega
Maxwell®
AS1220
PAAS1220EA 630.31 USD InStock
PAAS1220
Maxwell® 16 Tissue LEV Total RNA Purification Kit, Promega
Nucleic Acid Reagents Nucleic Acid Purification Kits and Reagents
The Maxwell 16 Tissue LEV Total RNA Purification Kit is used with the Maxwell 16 LEV Instrument.
  • For Automated Total RNA Purification from Mammalian Tissue and Whole Blood
  • Eluted RNA Concentration Is Ready for qRT-PCR for Most Sample Types
  • No Post-Purification Required
  • High Yields and High-Concentration Total RNA for Better Performance in Gene Expression Analysis Applications
  • Undetectable Contamination Genomic DNA Means Fewer Repeated Experiments and Unexplained or Variable Results
This RNA purification procedure provides an easy method for efficient, automated purification of highly concentrated total RNA from up to 25 mg of tissue and 2.5 mL of whole blood collected into PAXgene® tubes
No post-purification treatment with nuclease, cleanup, or concentration is required to achieve superior performance in downstream applications. The low elution volume (30–100 μL) is used to generate purified RNA in a more concentrated format for use in downstream applications such as qRT-PCR, RT-PCR, and cDNA synthesis.
Purified RNA for up to 16 samples is obtained in less than 45 minutes of hands-free instrument operation.
Accessories information: Maxwell 16 LEV Cartridge Rack Available Separately, One Each.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty