Would you like to log in again?
Chymotrypsin, Promega
Supplier: Promega
V1062
PAV1062EA 330 USD InStock
PAV1062
Chymotrypsin, Promega
Enzymes
Chymotrypsin is a highly-purified serine endopeptidase derived from bovine pancreas that preferentially hydrolyzes at the carboxyl side of aromatic amino acids: Tyr, Phe and Trp. Cleavage may also be observed, but at a lower rate, at Leu and Met.
  • Serine Protease that Cleaves at C-Terminus of Tyr, Phe and Trp
  • Optimal activity at pH 7.0-9.0.
  • Use alone or with other proteases for peptide mapping, peptide mass fingerprinting, MS/MS spectral matching
  • Use for digestion reactions in-solution or in-gel
Chymotrypsin is a highly-purified serine endopeptidase derived from bovine pancreas that preferentially hydrolyzes at the carboxyl side of aromatic amino acids: Tyr, Phe and Trp. Cleavage may also be observed, but at a lower rate, at Leu and Met. Chymotrypsin activity is optimal in the pH range of 7.0-9.0. This sequencing grade enzyme can be used alone or in combination with other proteases to produce protein digests for peptide mapping applications or protein identification by peptide mass fingerprinting or MS/MS spectral matching. It is suitable for digestion reactions in-solution or in-gel.
It is suitable for in-solution or in-gel digestion reactions.
Chymotrypsin activity is optimal in the pH range of 7.0–9.0. This sequencing-grade enzyme can be used alone or in combination with other proteases to produce protein digests for peptide mapping applications or protein identification by peptide mass fingerprinting or MS/MS spectral matching.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty