Would you like to log in again?
Grade
Supplier: Promega
V1621
PAV1621EA 248.52 USD InStock
PAV1621
Grade
Proteins and Peptides
Asp-N, Sequencing Grade, is an endoproteinase that hydrolyzes peptide bonds on the N-terminal side of aspartic and cysteic acid residues: Asp and Cys.
  • Hydrolyzes Peptide Bonds on the N-Terminal Side of Aspartic and Cysteic Acid Residues (Asp and Cys)
  • Optimal activity in pH range of 4-9
  • Sequencing grade enzyme for use alone or in combination with other proteases
  • Use for in-solution or in-gel digestion reactions
Asp-N, Sequencing Grade, is an endoproteinase that hydrolyzes peptide bonds on the N-terminal side of aspartic and cysteic acid residues: Asp and Cys. Asp-N activity is optimal in the pH range of 4.0-9.0. This sequencing grade enzyme can be used alone or in combination with other proteases to produce protein digests for peptide mapping applications or protein identification by peptide mass fingerprinting or MS/MS spectral matching. It is suitable for digestion reactions in solution or in gel.
It is suitable for digestion reactions in solution or in gel.
This sequencing-grade enzyme can be used alone or in combination with other proteases to produce protein digests for peptide mapping applications or protein identification by peptide mass fingerprinting or MS/MS spectral matching.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty