Would you like to log in again?
Glu-C
Supplier: Promega
V1651
PAV1651EA 232.37 USD InStock
PAV1651
Glu-C
Proteins and Peptides
Glu-C, Sequencing Grade (S. aureus V8), is a serine protease that specifically cleaves at the C-terminus of either aspartic or glutamic acid residues. Glu-C activity is optimal in the pH range of 4.0-9.0 and suitable for digestion reactions in solution.
  • Cleaves at the C-Terminus of Either Aspartic or Glutamic Acid Residues
  • Optimal activity at pH 4-9
  • Ammonium and phosphate buffers can affect cleavage; see details in product information
  • Suitable for in-solution digestion; not recommended for in-gel digestion
Glu-C, Sequencing Grade (S. aureus V8), is a serine protease that specifically cleaves at the C-terminus of either aspartic or glutamic acid residues. In ammonium bicarbonate and ammonium acetate the enzyme specificity is higher at the glutamic residues. In phosphate buffers cleavage occurs at the aspartic and glutamic residues. Glu-C activity is optimal in the pH range of 4.0-9.0. This sequencing grade enzyme can be used alone or in combination with other proteases to produce protein digests for peptide mapping applications or protein identification by peptide mass fingerprinting or MS/MS spectral matching. It is suitable for digestion reactions in solution.
This sequencing-grade enzyme can be used alone or in combination with other proteases to produce protein digests for peptide mapping applications or protein identification by peptide mass fingerprinting or MS/MS spectral matching. It is suitable for digestion reactions in solution but not recommended for in-gel digestions.
In phosphate buffers, cleavage occurs at the aspartic and glutamic residues. Glu-C activity is optimal in the pH range of 4.0–9.0.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty