Would you like to log in again?
rLys-C, Mass Spec Grade, 15microg, Promega
Supplier: Promega Corporation
V1671
PAV1671EA 193.33 USD InStock
PAV1671
rLys-C, Mass Spec Grade, 15microg, Promega
Proteins and Peptides
rLys-C, Mass Spec Grade, is a recombinant Lys-C expressed in E. coli. Sequence origin of rLys-C is Protease IV from Pseudomonas aeruginosa. Similar to a native Lys-C, rLys-C cleaves at the carboxyl side of lysine residues with exceptional specificity.
  • Recombinant Lys-C Cleaves at C-Terminus of Lysines
  • Optimal activity in the pH range of 8-9
  • Retains activity in denaturing conditions such as 8M urea
  • Available lyophilized or frozen
rLys-C, Mass Spec Grade, is a recombinant Lys-C expressed in E. coli. Sequence origin of rLys-C is Protease IV from Pseudomonas aeruginosa. Similar to a native Lys-C, rLys-C cleaves at the carboxyl side of lysine residues with exceptional specificity. rLys-C retains proteolytic activity under protein denaturing conditions such as 8M urea, which is used to improve digestion of proteolytically resistant proteins. rLys-C activity is optimal in the pH range of 8-9. The protease is supplied in a lyophilized form along with a Reconstitution Buffer, which is formulated to increase stability of rLys-C solution. Frozen rLys-C solution can be stored for a month at -20 degrees C without detectable loss of activity. rLys-C is recommended for digestion of single proteins and complex protein mixtures in-solution and in-gel.
Frozen rLys-C solution can be stored for a month at -20ºC without detectable loss of activity. rLys-C is recommended for digestion of in-solution and in-gel single proteins and complex protein mixtures.
Similar to a native Lys-C, rLys-C cleaves at the carboxyl side of lysine residues with exceptional specificity. rLys-C retains proteolytic activity under protein denaturing conditions such as 8M urea, which is used to improve digestion of proteolytically resistant proteins. rLys-C activity is optimal in the pH range of 8-9. The protease is supplied in a lyophilized form along with a Reconstitution Buffer, which is formulated to increase stability of rLys-C solution.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty