Would you like to log in again?
RNase A
Supplier: Promega
A7974
PAA7974EA 1452.08 USD InStock
PAA7974
RNase A
Enzymes
RNase A is an endoribonuclease that specifically hydrolyzes RNA 3´ of pyrimidine residues and cleaves the phosphodiester linkage to the adjacent nucleotide.
  • Endoribonuclease
  • For Removing RNA
  • Two Sizes Available
RNase A is used to remove RNA during procedures for the isolation of plasmid and genomic DNA.
The Solution is available in two sizes.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty